Assay Details
Gene Symbol: LOC705843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144470.2 | XP_014999956.1 | 525 | 95 | ||
XM_028851810.1 | XP_028707643.1 | 444 | 95 | |||
GenBank mRNA | JU327533.1 | - | 3 - 4 | 282 | 95 | |
JV531498.1 | - | 3 - 4 | 286 | 95 | ||
JV667329.1 | - | 3 - 4 | 292 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
