Assay Details
Gene Symbol: LOC705843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144470.2 | XP_014999956.1 | 823 | 64 | ||
XM_028851810.1 | XP_028707643.1 | 742 | 64 | |||
XM_028851811.1 | XP_028707644.1 | 586 | 64 | |||
GenBank mRNA | JU327533.1 | - | 6 - 7 | 580 | 64 | |
JV531498.1 | - | 6 - 7 | 584 | 64 | ||
JV667329.1 | - | 6 - 7 | 590 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
