Assay Details
Gene Symbol: LOC705843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144470.2 | XP_014999956.1 | 935 | 93 | ||
XM_028851810.1 | XP_028707643.1 | 854 | 93 | |||
XM_028851811.1 | XP_028707644.1 | 698 | 93 | |||
GenBank mRNA | JU327533.1 | - | 7 - 8 | 692 | 93 | |
JV531498.1 | - | 7 - 8 | 696 | 93 | ||
JV667329.1 | - | 7 - 8 | 702 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
