Assay Details
Gene Symbol: NUP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266179.1 | NP_001253108.1 | 539 | 126 | ||
GenBank mRNA | JU332685.1 | - | 516 | 126 | ||
JU474129.1 | - | 539 | 126 |

More Information
Set Membership: |
![]() ![]() |
