Assay Details
Gene Symbol: CASC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260566.1 | NP_001247495.1 | 7 - 8 | 1519 | 90 | |
XM_015119568.2 | XP_014975054.1 | 2732 | 90 | |||
GenBank mRNA | JU333443.1 | - | 7 - 8 | 1535 | 90 | |
JV046462.1 | - | 7 - 8 | 1519 | 90 | ||
JV609123.1 | - | 7 - 8 | 1542 | 90 | ||
JV636033.1 | - | 7 - 8 | 1525 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
