Assay Details
Gene Symbol: LOC114675369 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842628.1 | XP_028698461.1 | 441 | 154 |
Gene Symbol: TLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139468.2 | XP_014994954.1 | 2370 | 154 | ||
XM_028848870.1 | XP_028704703.1 | 2476 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() |
