Assay Details
Gene Symbol: DCAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098198.4 | XP_001098198.2 | 668 | 82 | ||
XM_015117537.2 | XP_014973023.2 | 553 | 82 | |||
XM_015117538.2 | XP_014973024.1 | 326 | 82 | |||
XM_015117539.2 | XP_014973025.1 | 384 | 82 | |||
GenBank mRNA | JU321756.1 | - | 544 | 82 | ||
JU471105.1 | - | 660 | 82 | |||
JU471106.1 | - | 660 | 82 | |||
JV045365.1 | - | 623 | 82 | |||
JV045366.1 | - | 623 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
