Assay Details
Gene Symbol: RDH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257659.1 | NP_001244588.1 | 6 - 7 | 894 | 119 | |
GenBank mRNA | JU334786.1 | - | 6 - 7 | 894 | 119 | |
JU334787.1 | - | 6 - 7 | 894 | 119 | ||
JU334788.1 | - | 6 - 7 | 894 | 119 | ||
JU334789.1 | - | 6 - 7 | 894 | 119 | ||
JU334790.1 | - | 6 - 7 | 894 | 119 | ||
JU475824.1 | - | 6 - 7 | 894 | 119 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
