Assay Details
Gene Symbol: CHRNA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001094288.4 | XP_001094288.1 | 3785 | 118 | ||
GenBank mRNA | JU489371.1 | - | 4 - 5 | 898 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
