Assay Details
Gene Symbol: KMT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028830088.1 | XP_028685921.1 | 1019 | 122 | ||
XM_028830090.1 | XP_028685923.1 | 515 | 122 | |||
XM_028830091.1 | XP_028685924.1 | 897 | 122 | |||
XM_028830092.1 | XP_028685925.1 | 722 | 122 | |||
GenBank mRNA | JU326980.1 | - | 4 - 5 | 536 | 122 | |
JU326981.1 | - | 4 - 5 | 536 | 122 | ||
JU326982.1 | - | 4 - 5 | 536 | 122 | ||
JU486277.1 | - | 4 - 5 | 589 | 122 | ||
JU486278.1 | - | 4 - 5 | 589 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
