Assay Details
Gene Symbol: ELOVL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040419.3 | NP_001035509.1 | 277 | 140 | ||
GenBank mRNA | JU322643.1 | - | 241 | 140 | ||
JU473227.1 | - | 399 | 140 | |||
JV566863.1 | - | 277 | 140 | |||
JV721926.1 | - | 272 | 140 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
