Assay Details
Gene Symbol: OCEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194595.2 | NP_001181524.2 | 2 - 3 | 287 | 131 | |
GenBank mRNA | JU325226.1 | - | 1 - 2 | 121 | 131 | |
JV481845.1 | - | 2 - 3 | 240 | 131 | ||
JV593627.1 | - | 2 - 3 | 230 | 131 | ||
JV628728.1 | - | 2 - 3 | 223 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() |
