Assay Details
Gene Symbol: IL20RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194658.3 | NP_001181587.2 | 652 | 86 | ||
XM_015132047.2 | XP_014987533.1 | 430 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
