Assay Details
Gene Symbol: PTCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001088008.4 | XP_001088008.1 | 1298 | 114 | ||
GenBank mRNA | JU331185.1 | - | 1 - 2 | 450 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
