Assay Details
Gene Symbol: RIMKLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261229.1 | NP_001248158.1 | 416 | 107 | ||
GenBank mRNA | JU334092.1 | - | 416 | 107 | ||
JU334093.1 | - | 416 | 107 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
