Assay Details
Gene Symbol: DTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152137.2 | XP_015007623.1 | 709 | 94 | ||
GenBank mRNA | JV046374.1 | - | 1 - 2 | 140 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015152137.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
