Assay Details
Gene Symbol: CENPV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118717.2 | XP_014974203.2 | 788 | 80 | ||
GenBank mRNA | JU474622.1 | - | 628 | 80 | ||
JV636015.1 | - | 655 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
