Assay Details
Gene Symbol: PRICKLE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260650.1 | NP_001247579.1 | 5 - 6 | 1073 | 106 | |
XM_015130445.2 | XP_014985931.1 | 787 | 106 | |||
XM_015130446.2 | XP_014985932.1 | 976 | 106 | |||
GenBank mRNA | JU333205.1 | - | 5 - 6 | 1073 | 106 | |
JV567187.1 | - | 5 - 6 | 1160 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
