Assay Details
Gene Symbol: LIPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028845411.1 | XP_028701244.1 | 744 | 117 | ||
XM_028845412.1 | XP_028701245.1 | 930 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
