Assay Details
Gene Symbol: CCDC187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116371.2 | XP_014971857.2 | 1262 | 137 | ||
XM_015116372.2 | XP_014971858.2 | 1264 | 137 | |||
XM_028834601.1 | XP_028690434.1 | 1264 | 137 |

More Information
Set Membership: |
![]() ![]() |
