Assay Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136019.2 | XP_014991505.2 | 704 | 144 | ||
XM_028835497.1 | XP_028691330.1 | 695 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
