Assay Details
Gene Symbol: RHBDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133303.2 | XP_014988789.1 | 392 | 146 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015133303.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
