Assay Details
Gene Symbol: CDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001096632.4 | XP_001096632.2 | 497 | 140 | ||
GenBank mRNA | JU328631.1 | - | 3 - 4 | 242 | 140 | |
JV666691.1 | - | 3 - 4 | 197 | 140 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
