Assay Details
Gene Symbol: DYRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152170.2 | XP_015007656.1 | 458 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015152170.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
