Assay Details
Gene Symbol: FKBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266723.1 | NP_001253652.1 | 3 - 4 | 337 | 108 | |
GenBank mRNA | JU320307.1 | - | 3 - 4 | 337 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
