Assay Details
Gene Symbol: GDF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146958.2 | XP_015002444.2 | 1724 | 66 | ||
GenBank mRNA | JV057822.1 | - | 1 - 2 | 372 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
