Assay Details
Gene Symbol: PHYHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266549.1 | NP_001253478.1 | 3 - 4 | 905 | 139 | |
GenBank mRNA | JU337332.1 | - | 3 - 4 | 905 | 139 | |
JV564374.1 | - | 3 - 4 | 915 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() |
