Assay Details
Gene Symbol: MYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151949.2 | XP_015007435.1 | 169 | 154 | ||
GenBank mRNA | JV684806.1 | - | 2 - 3 | 115 | 154 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015151949.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
