Assay Details
Gene Symbol: ARL6IP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257759.1 | NP_001244688.1 | 1 | 42 | 92 | |
GenBank mRNA | JU332530.1 | - | 1 | 42 | 92 | |
JV705324.1 | - | 1 | 65 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257759.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
