Assay Details
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139780.2 | XP_014995266.1 | 1216 | 120 | ||
GenBank mRNA | JV635779.1 | - | 5 | 1199 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
