Assay Details
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032929.1 | NP_001028101.1 | 1 - 2 | 128 | 80 | |
XM_028849023.1 | XP_028704856.1 | 1790 | 80 | |||
GenBank mRNA | AY244790.1 | - | 1 - 2 | 128 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
