Assay Details
Gene Symbol: LOC694631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082177.4 | XP_001082177.3 | 839 | 65 |
Gene Symbol: IFNA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194367.3 | NP_001181296.1 | 1 | 449 | 65 |
Gene Symbol: LOC710528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117656.2 | XP_014973142.2 | 386 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
