Assay Details
Gene Symbol: MIR10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032369.1 | 1 | 57 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ring finger and FYVE like domain containing E3 ubiquitin protein ligase | RFFL | Sense | Chr. 16 - 31387623 - 31468580 on NCBI Build Mmul_10 | ||
Downstream | RAD51 paralog D | RAD51D | 68.0kb | Sense | Chr. 16 - 31479272 - 31499640 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
