Assay Details
Gene Symbol: HSH2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190903.2 | NP_001177832.2 | 1 - 2 | 127 | 64 | |
XM_015123192.2 | XP_014978678.2 | 291 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
