Assay Details
Gene Symbol: GNB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190909.2 | NP_001177838.2 | 1 - 2 | 123 | 94 | |
XM_015149651.2 | XP_015005137.2 | 352 | 94 | |||
XM_028827449.1 | XP_028683282.1 | 555 | 94 | |||
XM_028827450.1 | XP_028683283.1 | 459 | 94 | |||
XM_028827451.1 | XP_028683284.1 | 403 | 94 | |||
GenBank mRNA | JU476095.1 | - | 3 - 4 | 424 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
