Assay Details
Gene Symbol: GNB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190909.2 | NP_001177838.2 | 4 - 5 | 516 | 90 | |
XM_015149651.2 | XP_015005137.2 | 745 | 90 | |||
XM_028827449.1 | XP_028683282.1 | 948 | 90 | |||
XM_028827450.1 | XP_028683283.1 | 852 | 90 | |||
XM_028827451.1 | XP_028683284.1 | 796 | 90 | |||
GenBank mRNA | JU328988.1 | - | 3 - 4 | 308 | 90 | |
JU476095.1 | - | 6 - 7 | 817 | 90 | ||
JV717368.1 | - | 3 - 4 | 353 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
