Assay Details
Gene Symbol: MIR887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032697.1 | 1 | 41 | 53 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | methionine adenosyltransferase 2B | MAT2B | -2.0mb | Anti-sense | Chr. 6 - 160864764 - 160881267 on NCBI Build Mmul_10 | |
Downstream | teneurin transmembrane protein 2 | TENM2 | ODZ2 | 1.5mb | Anti-sense | Chr. 6 - 164361139 - 165646202 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
