Assay Details
Gene Symbol: MIRLET7A-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032354.1 | 1 | 37 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | guanine nucleotide-binding protein G(s) subunit alpha | LOC694289 |
GNAS, GNAS1, TADA2B, XL |
-55.0kb | Sense | Chr. 10 - 93996247 - 94067999 on NCBI Build Mmul_10 |
Downstream | negative elongation factor complex member C/D | NELFCD | TH1L | 20.7kb | Sense | Chr. 10 - 94143747 - 94157496 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
