Assay Details
Gene Symbol: DDX46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261180.1 | NP_001248109.1 | 8 - 9 | 1159 | 91 | |
XM_015140860.2 | XP_014996346.1 | 1253 | 91 | |||
GenBank mRNA | JU337370.1 | - | 8 - 9 | 1190 | 91 | |
JU478067.1 | - | 8 - 9 | 1140 | 91 | ||
JV048852.1 | - | 8 - 9 | 1159 | 91 | ||
JV599884.1 | - | 8 - 9 | 1214 | 91 |
Gene Symbol: LOC107000127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015147052.2 | XP_015002538.2 | 790 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
