Assay Details
Gene Symbol: ISYNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261319.1 | NP_001248248.1 | 10 - 11 | 1531 | 94 | |
XM_015123293.2 | XP_014978779.2 | 1663 | 94 | |||
XM_028838525.1 | XP_028694358.1 | 2225 | 94 | |||
XM_028838526.1 | XP_028694359.1 | 2126 | 94 | |||
GenBank mRNA | JU334903.1 | - | 10 - 11 | 1547 | 94 | |
JU475908.1 | - | 10 - 11 | 1531 | 94 | ||
JV047308.1 | - | 10 - 11 | 1568 | 94 | ||
JV686389.1 | - | 10 - 11 | 1517 | 94 |
Gene Symbol: SSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV047307.1 | - | 10 - 11 | 1568 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
