Assay Details
Gene Symbol: ISYNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261319.1 | NP_001248248.1 | 2 - 3 | 177 | 109 | |
XM_028838526.1 | XP_028694359.1 | 677 | 109 | |||
GenBank mRNA | JU334903.1 | - | 2 - 3 | 193 | 109 | |
JU475908.1 | - | 2 - 3 | 177 | 109 | ||
JV047308.1 | - | 2 - 3 | 214 | 109 | ||
JV686389.1 | - | 2 - 3 | 163 | 109 |
Gene Symbol: SSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV047307.1 | - | 2 - 3 | 214 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
