Assay Details
Gene Symbol: ISYNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261319.1 | NP_001248248.1 | 3 - 4 | 345 | 70 | |
XM_015123293.2 | XP_014978779.2 | 477 | 70 | |||
XM_028838525.1 | XP_028694358.1 | 944 | 70 | |||
XM_028838526.1 | XP_028694359.1 | 845 | 70 | |||
GenBank mRNA | JU334903.1 | - | 3 - 4 | 361 | 70 | |
JU475908.1 | - | 3 - 4 | 345 | 70 | ||
JV047308.1 | - | 3 - 4 | 382 | 70 | ||
JV686389.1 | - | 3 - 4 | 331 | 70 |
Gene Symbol: SSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV047307.1 | - | 3 - 4 | 382 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
