Assay Details
Gene Symbol: ISYNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261319.1 | NP_001248248.1 | 4 - 5 | 477 | 98 | |
XM_015123293.2 | XP_014978779.2 | 609 | 98 | |||
GenBank mRNA | JU334903.1 | - | 4 - 5 | 493 | 98 | |
JU475908.1 | - | 4 - 5 | 477 | 98 | ||
JV047308.1 | - | 4 - 5 | 514 | 98 | ||
JV686389.1 | - | 4 - 5 | 463 | 98 |
Gene Symbol: SSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV047307.1 | - | 4 - 5 | 514 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
