Assay Details
Gene Symbol: ISYNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261319.1 | NP_001248248.1 | 9 - 10 | 1316 | 67 | |
XM_015123293.2 | XP_014978779.2 | 1448 | 67 | |||
XM_028838525.1 | XP_028694358.1 | 2010 | 67 | |||
XM_028838526.1 | XP_028694359.1 | 1911 | 67 | |||
GenBank mRNA | JU334903.1 | - | 9 - 10 | 1332 | 67 | |
JU475908.1 | - | 9 - 10 | 1316 | 67 | ||
JV047308.1 | - | 9 - 10 | 1353 | 67 | ||
JV686389.1 | - | 9 - 10 | 1302 | 67 |
Gene Symbol: SSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV047307.1 | - | 9 - 10 | 1353 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
