Assay Details
Gene Symbol: OSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267000.1 | NP_001253929.1 | 3 - 4 | 1102 | 111 | |
GenBank mRNA | JV635942.1 | - | 3 - 4 | 1102 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
