Assay Details
Gene Symbol: MPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260570.1 | NP_001247499.1 | 298 | 103 | ||
GenBank mRNA | JU322883.1 | - | 291 | 103 | ||
JU473816.1 | - | 232 | 103 | |||
JV044878.1 | - | 298 | 103 |

More Information
Set Membership: |
![]() ![]() |
