Assay Details
Gene Symbol: RHNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098483.4 | XP_001098483.3 | 87 | 78 | ||
XM_028829163.1 | XP_028684996.1 | 87 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001098483.4, XM_028829163.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
