Assay Details
Gene Symbol: LOC703700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145482.2 | XP_015000968.2 | 1651 | 94 | ||
XM_015145483.2 | XP_015000969.2 | 1674 | 94 | |||
XM_015145485.2 | XP_015000971.2 | 432 | 94 | |||
XM_015145486.2 | XP_015000972.2 | 551 | 94 | |||
XM_015145487.2 | XP_015000973.2 | 527 | 94 | |||
XM_015145488.2 | XP_015000974.2 | 709 | 94 | |||
XM_015145489.2 | XP_015000975.2 | 2101 | 94 | |||
XM_028852705.1 | XP_028708538.1 | 1200 | 94 | |||
XM_028852706.1 | XP_028708539.1 | 1166 | 94 | |||
XM_028852707.1 | XP_028708540.1 | 1972 | 94 | |||
XM_028852708.1 | XP_028708541.1 | 2100 | 94 | |||
XM_028852709.1 | XP_028708542.1 | 2101 | 94 | |||
XM_028852710.1 | XP_028708543.1 | 411 | 94 | |||
GenBank mRNA | JU320776.1 | - | 3 - 4 | 341 | 94 | |
JU320777.1 | - | 3 - 4 | 341 | 94 | ||
JV647133.1 | - | 3 - 4 | 342 | 94 | ||
JV711047.1 | - | 3 - 4 | 363 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
