Assay Details
Gene Symbol: NEURL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002804637.3 | XP_002804683.1 | 201 | 72 | ||
GenBank mRNA | JV548485.1 | - | 1 - 2 | 165 | 72 | |
JV600748.1 | - | 1 - 2 | 173 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002804637.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
