Assay Details
Gene Symbol: NEURL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002804637.3 | XP_002804683.1 | 1469 | 108 | ||
GenBank mRNA | JV600748.1 | - | 3 - 4 | 1441 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
